Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Phorbol-12-myristate-13-acetate
GSA_VA

Catalog No. 5005820001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-058-20001 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-058-20001 Supplier MilliporeSigma™ Supplier No. 5005820001
Add to Cart
Add to Cart

Most commonly-used phorbol ester

Most commonly-used phorbol ester. Extremely potent mouse skin tumor promoter. Activates protein kinase C in vivo and in vitro , even atnM concentrations. Promotes the expression of iNOS in cultured hepatocytes. Activates Ca2+-ATPase and potentiates forskolin (Cat. No. 344270, Cat. No. 344282)-induced cAMP formation. Inhibits apoptosis induced by the Fas antigen, but induces apoptosis in HL-60 promyelocytic leukemia cells. Its binding is reversible.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (2mg/324μL) solution of Phorbol-12-myristate-13-acetate in DMSO.
Description 98% by HPLC
Target Proteins PKC
Molecular Formula C36H56O8
Formula Weight 616.8g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.