Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ PI-103

Catalog No. 5281012MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-810-12MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-810-12MG Supplier MilliporeSigma™ Supplier No. 5281012MG
Add to Cart
Add to Cart

A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR (IC50 = 2, 8, 88, 48, 150, 26, 20, and 83nM for DNA-PK, p110α, p110β, p110δ, p110γ, PI3-KC2β, mTORC1, and mTORC2, respectively)

A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR (IC50 = 2, 8, 88, 48, 150, 26, 20, and 83nM for DNA-PK, p110α, p110β, p110δ, p110γ, PI3-KC2β, mTORC1, and mTORC2, respectively). It inhibits ATR and ATM only at much higher concentrations (IC50 = 850 and 920 nM, respectively) and exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10μM. Shown to effectively block PI3-K/Akt signaling and cell proliferation in glioma cell lines both in vitro and in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (2mg/574μl) solution of PI-103 in DMSO.
Description 97% by HPLC
Molecular Formula C19H16N4O3
Formula Weight 348.4g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.