Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ PKR Inhibitor

Catalog No. 5274515MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-745-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-745-15MG Supplier MilliporeSigma™ Supplier No. 5274515MG
Add to Cart
Add to Cart

The InSolution™ PKR Inhibitor, also referenced under CAS 608512-97-6, controls the biological activity of PKR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/373μL) solution of PKR Inhibitor in DMSO.
Description 90% by HPLC
Target Proteins PKR
CAS 608512-97-6
Formula Weight 268.3g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.