Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Rac1 Inhibitor II, Z62954982

Catalog No. 55351210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-351-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-351-210MG Supplier MilliporeSigma™ Supplier No. 55351210MG
Add to Cart
Add to Cart

The InSolution™ Rac1 Inhibitor II, Z62954982, also referenced under CAS 1090893-12-1, controls the biological activity of Rac1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 100 mM (10mg/241μl) solution of Rac1 Inhibitor II, Z62954982 in DMSO
Description 93% by HPLC
CAS 1090893-12-1
Molecular Formula C20H21N3O5S
Formula Weight 415.5g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.