Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Rac1 Inhibitor

Catalog No. 5535085MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-350-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-350-85MG Supplier MilliporeSigma™ Supplier No. 5535085MG
Add to Cart
Add to Cart

The InSolution™ Rac1Inhibitor, also referenced under CAS 1177865-17-6, controls the biological activity of Rac1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/185μl) solution of Rac1 Inhibitor in H2O.
Description 93% by HPLC
Target Proteins Rac1
Molecular Formula C24H35N7·3HCl
Formula Weight 531g/mol
Content And Storage Protect from light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.