Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Rho Kinase Inhibitor

Catalog No. 555552500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-555-2500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-555-2500UG Supplier MilliporeSigma™ Supplier No. 555552500UG
Add to Cart
Add to Cart

The InSolution™ Rho Kinase Inhibitor controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (500μg/128μl) solution of Rho Kinase Inhibitor in H2O.
Description 95% by HPLC
Target Proteins ROCK
Molecular Formula C16H21N3O2S·2HCl
Formula Weight 392.3g/mol
Content And Storage Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.