Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ RNA Polymerase III Inhibitor

Catalog No. 5574045MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-740-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-740-45MG Supplier MilliporeSigma™ Supplier No. 5574045MG
Add to Cart
Add to Cart

The InSolution™ RNA Polymerase III Inhibitor, also referenced under CAS 577784-91-9, controls the biological activity of RNA Polymerase III. This small molecule/inhibitor is primarily used for Cell Structure applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 25 mM (5mg/442μL) solution of RNA Polymerase III Inhibitor in DMSO.
Description 97% by HPLC
Target Proteins RNA Polymerase III
CAS 577784-91-9
Formula Weight 452.4g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.