Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Roscovitine

Catalog No. 5573645MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-736-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-736-45MG Supplier MilliporeSigma™ Supplier No. 5573645MG
Add to Cart
Add to Cart

A potent and selective inhibitor of cyclin-dependent kinases (Cdks)

A potent and selective inhibitor of cyclin-dependent kinases (Cdks). Exhibits greater inhibitory potency compared to olomoucine (Cat. No. 495620). Inhibits p34 Cdc2/cyclin B (IC50 = 650nM), p33 Cdk2/cyclin A (IC50 = 700nM), p33 Cdk2/cyclin E (IC50 = 700nM), and p33 Cdk2/p35 (IC50 = 200nM) by competing for the ATP binding domain of these kinases. Exhibits very low sensitivity towards related kinases, such as Erk1 and Erk2 (IC50 = 34μM and 14μM, respectively). Roscovitine does not significantly affect the activity of other protein kinases even at 100μM and compared to olomoucine, roscovitine displays increased anti-mitotic activity at the G1/S and G2/M phases of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/282μl) solution of Roscovitine in DMSO.
Description 95% by HPLC
Target Proteins p34cdk1/cyclin B
Molecular Formula C19H26N6O
Formula Weight 354.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.