Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ SB 203580

Catalog No. 5593981ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-939-81ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-939-81ML Supplier MilliporeSigma™ Supplier No. 5593981ML
Add to Cart
Add to Cart

InSolution SB 203580, CAS 152121-47-6, is a 1mg/mL solution of SB 203580 in DMSO. A cell-permeable, selective, reversible inhibitor of p38 MAP kinase (IC50 = 34nM in vitro, 600nM in cells).

InSolution SB 203580, CAS 152121-47-6, is a 1mg/mL solution of SB 203580 in DMSO. A cell-permeable, selective, reversible inhibitor of p38 MAP kinase (IC50 = 34nM in vitro, 600nM in cells).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 1mg/mL solution of SB 203580 in anhydrous DMSO.
Description 98% by HPLC
Target Proteins P38MAPK
Molecular Formula C21H16FN3OS
Formula Weight 377.4g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.