Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Sirtinol

Catalog No. 5663211MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-632-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-632-11MG Supplier MilliporeSigma™ Supplier No. 5663211MG
Add to Cart
Add to Cart

A cell-permeable 2-hydroxy-1-napthaldehyde derivative that acts as a specific and a direct inhibitor of sirtuin class of histone deacetylase (HDAC) activity

A cell-permeable 2-hydroxy-1-napthaldehyde derivative that acts as a specific and a direct inhibitor of sirtuin class of histone deacetylase (HDAC) activity. Does not affect human HDAC1. Reported to block Sir2p transcriptional silencing activity in vivo (IC50 = 25μM) and NAD-dependent HDAC activity in purified recombinant yeast Sir2p and human SIRT2 in vitro (IC50 = 68μM and 38μM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg/254μl) solution of Sirtinol in DMSO.
Description 97% by HPLC
Target Proteins ySir2
Molecular Formula C26H22N2O2
Formula Weight 394.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.