Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ STAT3 Inhibitor III, WP1066

Catalog No. 5731295MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-312-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-312-95MG Supplier MilliporeSigma™ Supplier No. 5731295MG
Add to Cart
Add to Cart

The InSolution™ STAT3 Inhibitor III, WP1066, also referenced under CAS 857064-38-1, controls the biological activity of STAT3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/281μl) solution of STAT3 Inhibitor III, WP1066 in DMSO.
Description 97% by HPLC
Molecular Formula C17H14BrN3O
Formula Weight 356.2g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.