Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ SU5402

Catalog No. 572631500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-263-1500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-263-1500UG Supplier MilliporeSigma™ Supplier No. 572631500UG
Add to Cart
Add to Cart

Inhibits the tyrosine kinase activity of FGFR1 (IC50 = 10-20μM in the presence of 1mm ATP) by interacting with the catalytic domain of FGFR1

Inhibits the tyrosine kinase activity of FGFR1 (IC50 = 10-20μM in the presence of 1mm ATP) by interacting with the catalytic domain of FGFR1. Also inhibits aFGF-induced phosphorylation of ERK1 and ERK2 (IC50 = 10-20μM). In contrast to SU4984, it acts only as a weak inhibitor of tyrosine phosphorylation of the PDGF receptor. Does not inhibit the phosphorylation of insulin receptor and exhibits no inhibitory effect on EGF receptor kinase.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (500μg/169μl) solution of SU5402 in DMSO.
Description 95% by HPLC
Molecular Formula C17H16N2O3
Formula Weight 296.3g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.