Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ SU6656

Catalog No. 572636500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-263-6500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-263-6500UG Supplier MilliporeSigma™ Supplier No. 572636500UG
Add to Cart
Add to Cart

A potent inhibitor of Src-family protein tyrosine kinases

A potent inhibitor of Src-family protein tyrosine kinases. Inhibits Src (IC50 = 280nM) as well as closely related kinases Fyn (IC50 = 170nM), Yes (IC50 = 20nM) and Lyn (IC50 = 130nM). The inhibition is competitive with respect to ATP. Has only a trivial effect on Lck (IC50 = 6.88μM). Does not affect the activity of PDGF-β receptor kinase (IC50 >10μM) and IGF-1 receptor kinase (IC50 gt;20μM).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (500μg/135μl) solution of SU6656 in DMSO.
Description 95% by HPLC
Target Proteins Src
Molecular Formula C19H21N3O3S
Formula Weight 371.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.