Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ TAPI-1

Catalog No. 579053500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-905-3500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-905-3500UG Supplier MilliporeSigma™ Supplier No. 579053500UG
Add to Cart
Add to Cart

InSolution™ TAPI-1, CAS 171235-71-5, is a 10mm solution of TAPI-1 in DMSO. A structural analog of TAPI-0 (Cat. No. 579050) with similar in vitro efficacy for the inhibition of MMPs and TACE.

InSolution™ TAPI-1, CAS 171235-71-5, is a 10mm solution of TAPI-1 in DMSO. A structural analog of TAPI-0 (Cat. No. 579050) with similar in vitro efficacy for the inhibition of MMPs and TACE.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (500μg/100μl) solution of TAPI-1 in DMSO.
Description 97% by HPLC
Target Proteins MMPs and TACE
Molecular Formula C26H37N5O5
Formula Weight 499.6g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.