Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Tetrahydrouridine

Catalog No. 58422325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-422-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58-422-325MG Supplier MilliporeSigma™ Supplier No. 58422325MG
Add to Cart
Add to Cart

Tetrahydrouridine is a potent competitive inhibitor of cytidine deaminase.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 100 mM (25mg/879μL) solution of Tetrahydrouridine in H2O.
Description 85% by HPLC
Target Proteins cytidine deaminase
CAS 18771-50-1
Formula Weight 284.3g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.