Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ TGF-β RI Kinase Inhibitor II, RepSox

Catalog No. 5081580001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-815-80001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-815-80001 Supplier MilliporeSigma™ Supplier No. 5081580001
Add to Cart
Add to Cart

A cell-permeable, potent, selective, reversible, and ATP-competitive inhibitor of TGF-β type I receptor (ALK5; IC50 = 23 nM, 4nM and 18nM for ALK5 binding, ALK5 auto-phosphorylation, and TGF-β cellular assay in HepG2 cells, respectively)

A cell-permeable, potent, selective, reversible, and ATP-competitive inhibitor of TGF-β type I receptor (ALK5; IC50 = 23 nM, 4nM and 18nM for ALK5 binding, ALK5 auto-phosphorylation, and TGF-β cellular assay in HepG2 cells, respectively). Minimally affects a panel of 9 closely related kinases including p38 MAPK (IC50 >16μM).
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 50 mM (5mg/348μl) sterile-filtered solution of TGF-β RI Kinase Inhibitor II, RepSox in DMSO.
Description 99% by HPLC
Target Proteins ALK5
Molecular Formula C17H13N5
Formula Weight 287.3g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.