Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Trichostatin A, Streptomyces sp.

Catalog No. 647926500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-792-6500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-792-6500UG Supplier MilliporeSigma™ Supplier No. 647926500UG
Add to Cart
Add to Cart

A potent and reversible inhibitor of histone deacetylase

A potent and reversible inhibitor of histone deacetylase. Blocks cell cycle progression at the G1 phase in HeLa cells and induces a 12-fold increase in intracellular levels of gelsolin. Induces reversion of oncogenic ras-tranformed NIH 3T3 cells to a normal morphology. Inhibits IL-2 gene expression (IC50 = 73nM) in Jurkat cells and shows immunosuppressive activity in a mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (500μg/165μl) solution of Trichostatin A, Streptomyces sp. in DMSO.
Description 98% by HPLC
Target Proteins histone deactylase
Molecular Formula C17H22N2O3
Formula Weight 302.4g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.