Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Vitamin D3, 1α, 25-Dihydroxy-

Catalog No. 5097210001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-972-10001 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-972-10001 Supplier MilliporeSigma™ Supplier No. 5097210001
Add to Cart
Add to Cart

A 1mm (50 ug/112μL) solution of Vitamin D3, 1α, 25-Dihydroxy- in DMSO

Active hormonal form of vitamin D, formed by the renal 1-hydroxylation of 25-(OH)D3. Has immunomodulatory properties in vitro and in vivo . Prevents the development of clinical diabetes in NOD mice, which are an animal model of human autoimmune diabetes. Induces reversible hydrolysis of the sphingomyelin cycle and generates ceramide. Stimulates intestinal Ca2+ and phosphate transport and mobilizes Ca2+ from bone. Involved in cellular differentiation, carcinogenesis, and insulin secretion.
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 1 mM (50 ug/112μL) solution of Vitamin D3, 1α, 25-Dihydroxy- in DMSO.
Description 99% by HPLC
Target Proteins vitamin D receptor
Molecular Formula C27H44O3
Formula Weight 416.7g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light
Form Liquid
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.