Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ Wortmannin

Catalog No. 6816761MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-167-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-167-61MG Supplier MilliporeSigma™ Supplier No. 6816761MG
Add to Cart
Add to Cart

Wortmannin is a fungal metabolite that acts as a potent, selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Formulation A 10 mM (1mg/233μl) solution of Wortmannin in DMSO.
Description 95% by HPLC
Molecular Formula C23H24O8
Formula Weight 428.4g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.