Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Interleukin-1 Receptor-Associated-Kinase-1/4 Inhibitor

Catalog No. 4076015MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-760-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-760-15MG Supplier MilliporeSigma™ Supplier No. 4076015MG
Add to Cart
Add to Cart

The Interleukin-1 Receptor-Associated-Kinase-1/4 Inhibitor controls the activity of Interleukin-1 Receptor-Associated-Kinase-1/4. Primarily used for Phosphorylation & Dephosphorylation apps.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Target Proteins Interleukin-1 receptor-associated kinases
CAS 509093-47-4
Molecular Formula C20H21N5O4
Formula Weight 395.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Crystalline Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.