Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ iPSC Induction Enhancer, Thiazovivin

Catalog No. 42022010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-022-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-022-010MG Supplier MilliporeSigma™ Supplier No. 42022010MG
Add to Cart
Add to Cart

A cell-permeable aminothiazolo-carboxamide compound that is reported to augment SB431542 and PD0325901.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 98% by HPLC
Molecular Formula C15H13N5OS
Formula Weight 311.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.