Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IRAK4 Inhibitor, Compound 26

Catalog No. 5312370001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-123-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-123-70001 Supplier MilliporeSigma™ Supplier No. 5312370001
Add to Cart
Add to Cart

A cell permeable indolo[2,3-c]quinoline derivatived compound that acts as a selective and highly potent inhibitor of interleukin-1 receptor associated kinase (IRAK4; IC50 = 94

A cell permeable indolo[2,3-c]quinoline derivatived compound that acts as a selective and highly potent inhibitor of interleukin-1 receptor associated kinase (IRAK4; IC50 = 94 pM). Displays much reduced potency against IRAK1 (IC50 = 65nM) and displays high selectivity over a panel of other 27 kinases, including several involved in TLR-mediated signaling. Blocks R848-induced TNF-α and IL-6 production in primary human monocytes and inhibits LPS-induced TNF-α production in mice (100 mpk). Shows minimal inhibition of Cyp2C9, 2D6, and 3A4 and has excellent microsomal stability and desirable aqueous solubility (100mg/mL at pH 7.4).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins IRAK4
Molecular Formula C25H27N5O2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.