Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IRE1 Inhibitor I, STF-083010

Catalog No. 41251010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-251-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-251-010MG Supplier MilliporeSigma™ Supplier No. 41251010MG
Add to Cart
Add to Cart

The IRE1 Inhibitor I, STF-083010 controls the biological activity of IRE1. This small molecule/inhibitor is primarily used for Biochemicals applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 98% by HPLC
Molecular Formula C15H11NO3S2
Formula Weight 317.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.