Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ischemin

Catalog No. 41096025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-096-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-096-025MG Supplier MilliporeSigma™ Supplier No. 41096025MG
Add to Cart
Add to Cart

A cell-permeable azobenzene compound that reversibly targets CBP-BRD acetyl-lysine binding pocket with moderate selectivity over BAZ1B, PCAF, BRD4-1 and BAZ2B.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-red
Description 99% by HPLC
Molecular Formula C15H17N3SO4
Formula Weight 335.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.