Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ ITK Inhibitor, BMS-509744

Catalog No. 4198205MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-982-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-982-05MG Supplier MilliporeSigma™ Supplier No. 4198205MG
Add to Cart
Add to Cart

A cell-permeable aminothioaryl-thiazolo compound that potently inhibits ITK kinase activity (IC50 = 19nM; [ATP] = 1μM) in an ATP-competitive manner by stabilizing ITK activation loop in a substrate-blocking, inactive conformation, inhibiting Fyn, IR, Lck, Btk only at much higher concentrations (IC50

A cell-permeable aminothioaryl-thiazolo compound that potently inhibits ITK kinase activity (IC50 = 19nM; [ATP] = 1μM) in an ATP-competitive manner by stabilizing ITK activation loop in a substrate-blocking, inactive conformation, inhibiting Fyn, IR, Lck, Btk only at much higher concentrations (IC50 ≥1.1μM) and exhibiting little or no activity against 14 other kinases (IC50 ≥11μM). Inhibits ITK-dependent cellular signaling (IC50 <300nM against αCD3-stimulated PLCγ1 Tyr phosphorylation and Ca2+ mobilization in Jurkat cells) in vitro and alleviate OVA (Cat. No. 32467) challenge-induced airway leukocytes infiltration (ED50 = 25mg/kg s.c.) in OVA-sensitized mice in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 97% by HPLC
Target Proteins ITK
CAS 439575-02-7
Molecular Formula C32H41N5O4S2
Formula Weight 623.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.