Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ JAK2 Inhibitor III, SD-1029

Catalog No. 57309810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-309-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-309-810MG Supplier MilliporeSigma™ Supplier No. 57309810MG
Add to Cart
Add to Cart

The JAK2 Inhibitor III, SD-1029, also referenced under CAS 118372-34-2, controls the biological activity of JAK2.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 80% by HPLC (sum of isomers)
CAS 118372-34-2
Molecular Formula C25H30Cl2N2O3·2HBr
Formula Weight 639.3g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.