Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Jasplakinolide, Jaspis johnstoni

Catalog No. 42010750UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-010-750UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-010-750UG Supplier MilliporeSigma™ Supplier No. 42010750UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable F-actin probe.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins Inducer of actin polymerization and stabilization <i>in vitro</i>
CAS 102396-24-7
Molecular Formula C36H45BrN4O6
Formula Weight 709.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.