Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ JNK Inhibitor X, BI-78D3

Catalog No. 4201405MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-014-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-014-05MG Supplier MilliporeSigma™ Supplier No. 4201405MG

The JNK Inhibitor X, BI-78D3, also referenced under CAS 883065-90-5, controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
CAS 883065-90-5
Molecular Formula C13H9N5O5S2
Formula Weight 379.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.