Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ K-252c

Catalog No. 4203051MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-030-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-030-51MG Supplier MilliporeSigma™ Supplier No. 4203051MG
Add to Cart
Add to Cart

A cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (IC50 = 2.45 μM) and protein kinase A (IC50 = 25.7 μM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins PKC
CAS 85753-43-1
Molecular Formula C20H13N3O
Formula Weight 311.3g/mol
Solubility DMSO or MeOH
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.