Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Kainic Acid

Catalog No. 42031810MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-031-810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-031-810MG Supplier MilliporeSigma™ Supplier No. 42031810MG
Add to Cart
Add to Cart

An excitatory amino acid receptor agonist selective for the kainate receptor subtype (Ki = 21nM for 3H-kainate binding in rat striatum).

An excitatory amino acid receptor agonist selective for the kainate receptor subtype (Ki = 21nM for 3H-kainate binding in rat striatum). Also activates AMPA receptors.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins Kainate receptor subtype
CAS 487-79-6
Molecular Formula C10H15NO4
Formula Weight 213.2g/mol
Solubility H2O or Dilute Alkali
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.