Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Kallikrein Substrate, Fluorogenic

Catalog No. 4203145MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-031-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-031-45MG Supplier MilliporeSigma™ Supplier No. 4203145MG
Add to Cart
Add to Cart

Sensitive fluorogenic substrate for kallikreins.

Peptide Sequence: H-D-Val-Leu-Arg-AFC, 2HCl
TRUSTED_SUSTAINABILITY

Specifications

Description 95% by HPLC
Target Proteins Fluorogenic substrate for kallikreins
Molecular Formula C27H38N7O5F3·2HCl
Formula Weight 670.6g/mol
Solubility DMSO or Acetic Acid
Content And Storage Protect from moisture
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.