Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ketanserin Tartrate

Catalog No. 5062710001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-627-10001 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-627-10001 Supplier MilliporeSigma™ Supplier No. 5062710001
Add to Cart
Add to Cart

A selective 5-HT2A serotonin receptor antagonist with weak adrenergic receptor blocking propert

A selective 5-HT2A serotonin receptor antagonist with weak adrenergic receptor blocking properties. Strong affinity for 5-HT2AR (Ki = 1.49nM) relative to other family members like 5-HT2BR (Ki = 740nM) and 5-HT2CR (Ki = 59nM). Often used in conjunction with SB 206553 to distinguish 5-HT2AR and 5-HT2CR activity.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins 5-HT2A
Molecular Formula C22H22FN3O3·C4H6O6
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.