Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ KN-92

Catalog No. 4227091MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-270-91MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-270-91MG Supplier MilliporeSigma™ Supplier No. 4227091MG
Add to Cart
Add to Cart

KN-92, CAS 1135280-28-2, is useful as a negative control for KN-93 (Cat. No. 422708), a Ca2+/CaM Kinase II inhibitor.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Useful as a negative control for KN-93 (Cat. No. 422708), a Ca2+/CaM kinase II inhibitor
CAS 1135280-28-2
Molecular Formula C24H25ClN2O3S·H3PO4
Formula Weight 555g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.