Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ L-Ascorbic Acid, Free Acid

Catalog No. 18311KG Shop All MilliporeSigma Products
Change view
Click to view available options
L-Ascorbic Acid, Free Acid; Qty: 1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-311-KG L-Ascorbic Acid, Free Acid; Qty: 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-311-KG Supplier MilliporeSigma™ Supplier No. 18311KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects cells against damaging effects of radiation and oxygen radicals

  • Increases rate of mineralization in osteoblasts
  • Reduces Na+, Ca2+ exchange in cultured astrocytes
  • Modulates cyclic nucleotide levels in B and T cells
  • Inhibits apoptosis in cultured rat ovarian follicles

Note: Material readily oxidizes in aqueous solution; reconstitute just prior to use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Additive L-Ascorbic Acid, Free Acid
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.