Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ L- Buthionine Sulfoximine

Catalog No. 5082280001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-822-80001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-822-80001 Supplier MilliporeSigma™ Supplier No. 5082280001
Add to Cart
Add to Cart

A cell-permeable, sulfoximine based compound that acts a potent, fast acting, irreversible inhibitor of g-glutamylcysteine synthetase and depletes cellular glutathione levels

A cell-permeable, sulfoximine based compound that acts a potent, fast acting, irreversible inhibitor of g-glutamylcysteine synthetase and depletes cellular glutathione levels. Closely resembles the structure of g-glutamylphosphate cysteine adduct. The inhibition follows the first order kinetics with t1/2 = ∽ 11 sec at saturating inhibitor concentration. Does not affect the activity of glutamine synthetase. Shown to improve the outcome of chemotherapy by lowering the levels of glutathione in cancer cells (IC50 = 1.9, 8.6, and 29μM in melanoma, breast, and ovarian tumor cells, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by TLC
Target Proteins gamma-glutamylcysteine synthetase
Molecular Formula C8H18N2O3S
Solubility H2O
Content And Storage Protect from light
Form Powder
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.