Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ L-Serine-O-phosphate

Catalog No. 5097050001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-970-50001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-970-50001 Supplier MilliporeSigma™ Supplier No. 5.09705.0001

missing translation for 'validMainframeMsg'

A potent agonist for group III mGlu receptor subtype 4, 6, and 8.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins mGlu4, mGlu6, mGlu8
CAS 407-41-0
Molecular Formula C3H8NO6P
Solubility H2O
Content And Storage Protect from light
Form Powder
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.