Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ L3MBTL1 Ligand, UNC669

Catalog No. 44264025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-264-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-264-025MG Supplier MilliporeSigma™ Supplier No. 44264025MG
Add to Cart
Add to Cart

A nicotinamido-pyrrolidine compound that targets the second MBT (Malignant Brain Tumor) module of chromatin lock L3MBTL1.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Orange-yellow
Description 95% by HPLC
Molecular Formula C15H20BrN3O
Formula Weight 338.3g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.