Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Latrunculin A, Latrunculia magnifica

Catalog No. 428021100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-802-1100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-802-1100UG Supplier MilliporeSigma™ Supplier No. 428021100UG
Add to Cart
Add to Cart

Latrunculin A, CAS 76343-93-6, is a cell-permeable marine toxin that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (KD = 200 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins G-actin
CAS 76343-93-6
Molecular Formula C22H31NO5S
Formula Weight 421.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.