Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LDN-211904

Catalog No. 4282015MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-820-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-820-15MG Supplier MilliporeSigma™ Supplier No. 4282015MG
Add to Cart
Add to Cart

LDN-211904 modulates the biological activity of EphB3 receptor tyrosine kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins Erythropoietin-producing hepatocellular carcinoma (Eph)
Molecular Formula C21H21ClN4O5
Formula Weight 444.87g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.