Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Leukadherin-1

Catalog No. 43461025MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-461-025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-461-025MG Supplier MilliporeSigma™ Supplier No. 43461025MG
Add to Cart
Add to Cart

A furanyl thiazolidinone compound that binds CD11b αA/αI domain in a reversible manner and stabilizes αA in a high-affinity binding conformation toward known CD11b/CD18 (αMβ2; CR3; Mac-1) ligands, fibrinogen, ICAM-1, and iC3B.

A furanyl thiazolidinone compound that binds CD11b αA/αI domain in a reversible manner and stabilizes αA in a high-affinity binding conformation toward known CD11b/CD18 (αMβ2; CR3; Mac-1) ligands, fibrinogen, ICAM-1, and iC3B. Reported to be superior to IMB-10 (Cat. No. 407271) in enhancing Mac-1-mediated cell adhesion (EC50 = 4 vs. >50μM on fibrinogen surface). Effectively suppresses leukocyte recruitment in various inflammatory disease models involving mice, rats, and zebrafish embryos in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color Red-brown
Description 95% by HPLC
Molecular Formula C22H15NO4S2
Formula Weight 421.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.