Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Licochalcone-A, Synthetic
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Yellow |
| Target Proteins | Anti-tumor and antiparasitic properties |
| CAS | 58749-22-7 |
| Molecular Formula | C21H22O4 |
| Formula Weight | 338.4g/mol |
| Solubility | DMSO or EtOH |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 50 mg |
| Purity | 96% by HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction