Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LipidGreen2

Catalog No. 5060290001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-602-90001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-602-90001 Supplier MilliporeSigma™ Supplier No. 5060290001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable stain for neutral lipids

  • A cell-permeable N-dimethylallylated 3-hydroxyindole based compound that is shown to selectively stain neutral lipids, but not phospholipids, in various cells lines with bright fluorescence and with minimal non-specific background
  • Generates stronger staining with lipid droplets (∼10μM) when compared to Nile Red
  • Shown to be more effective than BODIPY™ 493/503 in assessing hepatotoxicity of several known compounds using HepG2 cells
  • Also suitable for staining fat deposits in developing zebrafish embryos
  • Can be used in conjunction with LipidToxt red dye for multicolor imaging of neutral lipids
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-yellow
Content And Storage Store at −20°C
Molecular Formula C22H29NO4
Packaging Type Packaged under inert gas
Purity ≥97% by HPLC
Quantity 5 mg
Form Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.