Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Loperamide Hydrochloride

Catalog No. 5081620001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-816-20001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-816-20001 Supplier MilliporeSigma™ Supplier No. 5081620001
Add to Cart
Add to Cart

A very potent and highly selective peripheral resctrictedμ-opioid receptor agonist (Ki = 2

A very potent and highly selective peripheral resctrictedμ-opioid receptor agonist (Ki = 2nM). Often used as an antidiarrheal, antipruritic, and antihyperalgesic agent. In addition, shown to improve defective post-receptor insulin signaling in skeletal muscle cells and enhance insulin sensitivity in rats.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins u opioid receptor
Molecular Formula C29H33ClN2O2·HCl
Solubility DMSO or Warm EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.