Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Lovastatin, Sodium Salt

Catalog No. 4381865MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-818-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-818-65MG Supplier MilliporeSigma™ Supplier No. 4381865MG
Add to Cart
Add to Cart

Carboxylate form of Lovastatin that is active in whole cells and cell-free assays.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase
Molecular Formula C24H37O6·Na
Formula Weight 444.5g/mol
Solubility DMSO or EtOH
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.