Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Lovastatin

Catalog No. 43818525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-818-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-818-525MG Supplier MilliporeSigma™ Supplier No. 43818525MG
Add to Cart
Add to Cart

Lovastatin, CAS 75330-75-5, is an anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase.

An anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase. Has been shown to block a series of biological events including: the activation of p21 ras by insulin in 3T3-L1 fibroblasts and 3T3-L1 adipocytes; the farnesylation of p21 ras , which decreases the pool of intracellular Ras available for subsequent activation by growth factors (including insulin); and N- ras -induced neuronal differentiation. Causes cell cycle arrest in the late G1 phase through inhibition of proteasome. Has recently been shown to inhibit the stimulation of MAP kinase by insulin in HIRcB cells and to block the transcription of type-I SCR gene in THP-1-derived macrophages. Also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF. This product requires activation for use in cell synchronization studies and in cell-free assay systems for HMG-CoA studies.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Description 95% by HPLC
Target Proteins Activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase
CAS 75330-75-5
Molecular Formula C24H36O5
Formula Weight 404.5g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Powder
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.