Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LSD1 Inhibitor II, S2101

Catalog No. 4894775MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-947-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-947-75MG Supplier MilliporeSigma™ Supplier No. 4894775MG
Add to Cart
Add to Cart

The LSD1 Inhibitor II, S2101 controls the biological activity of LSD1. This small molecule/inhibitor is primarily used for Cell Signaling applications.

A cell-permeable, 2-PCPA derivative that elicits mechanism-based inhibition against LSD1 (IC50 = 0.99μM, KI = 0.61μM, and kinact/KI = 4560 M-1S-1), and displays much lower inhibition activity toward MAO-B (KI = 17μM, kinact/KI = 18 M-1S-1) and MAO-A (KI = 110μM, kinact/KI = 60 M-1S-1), with higher potency and selectivity compared with those of 2-PCPA (Cat. No. 616431) (IC50 = 184μM, KI = 100μM, kinact/KI = 58 M-1S-1 for LSD1, and K I = 26μM, kinact/KI = 271 M-1S-1 for MAO-B, and K I = 5μM, kinact/KI = 1050 M-1S-1 for MAO-A). The inhibition of LSD1 is further confirmed in a cell-based assay, the treatment of HEK293T human cells with S2101 results in a dose-dependent increase in the level of H3K4me2, and about 50-fold stronger inhibition compared with that of 2-PCPA.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Molecular Formula C16H16ClF2NO
Formula Weight 311.75g/mol
Solubility DMSO or H2O
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.