Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LSD1 Inhibitor IV, RN-1,HCl

Catalog No. 48947910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-947-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-947-910MG Supplier MilliporeSigma™ Supplier No. 48947910MG
Add to Cart
Add to Cart

LSD1 Inhibitor IV, RN-1, HCl, is a cell-permeable potent, irreversible inhibitor of lysine specific demethylase 1 (LSD1; IC50 = 70 nM in a HRP-coupled assay using H3K4Me2 peptide substrate).

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by HPLC
Molecular Formula C23H29N3O2·HCl
Formula Weight 416g/mol
Solubility H2O or DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.