Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ LSF Inhibitor, FQI1

Catalog No. 43821010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-821-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-821-010MG Supplier MilliporeSigma™ Supplier No. 43821010MG
Add to Cart
Add to Cart

The LSF Inhibitor, FQI1 controls the biological activity of LSF.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >92% by HPLC
Molecular Formula C18H17NO4
Formula Weight 311.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.