Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glycogen Phosphorylase Inhibitor

Catalog No. 3615155MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-151-55MG 5 mg
36-151-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-151-55MG Supplier MilliporeSigma™ Supplier No. 3615155MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Glycogen Phosphorylase Inhibitor, also referenced under CAS 648926-15-2, controls the biological activity of Glycogen Phosphorylase.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins Glycogen phosphorylase
Molecular Formula C17H15ClF2N4O4
Formula Weight 412.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.