Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GSK-3 Inhibitor IX

Catalog No. 36155030MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
30 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-155-030MG 30 mg
36-155-01MG 1 mg
36-155-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 36-155-030MG Supplier MilliporeSigma™ Supplier No. 36155030MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BIO, CAS 667463-62-9, is a cell-permeable, highly potent, selective, reversible, and ATP-competitive inhibitor of GSK-3α/β (IC50 = 5 nM). Maintains self-renewal in human & mouse embryonic stem cells.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Purple
Product Type Inhibitor
Target Proteins GSK-3α/β
CAS 667463-62-9
Molecular Formula C16H10BrN3O2
Formula Weight 356.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 30 mg
Purity 97% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.